LIF372153089 Life Chemicals Inc.

N-Methyl-N-[4-(morpholin-4-ylsulfonyl)benzoyl]glycine

  • MDL number MFCD03723713
  • Cas Number : 620103-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart