LIF372153090 Life Chemicals Inc.

1-{[(4-Chlorophenyl)sulfonyl]amino}cyclohexanecarboxylic acid

  • MDL number MFCD02081088
  • Cas Number : 314744-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart