LIF372153093 Life Chemicals Inc.

2-(3-methoxyphenoxy)acetic acid

  • MDL number MFCD00014357
  • Cas Number : 2088-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart