LIF372153119 Life Chemicals Inc.

4-[(4-acetylpiperazin-1-yl)carbonyl]benzoic acid

  • MDL number MFCD05862408
  • Cas Number : 896508-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart