LIF372153120 Life Chemicals Inc.

2-{[4-(2-Chlorobenzoyl)piperazin-1-yl]carbonyl}benzoic acid

  • MDL number MFCD05862417
  • Cas Number : 896508-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart