LIF372153126 Life Chemicals Inc.

7,7-Dimethyl-2,3-dioxo-bicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD05859737
  • Cas Number : 139492-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart