LIF372153140 Life Chemicals Inc.

7-allyl-6-hydroxy-2-phenyl-4H-chromen-4-one

  • MDL number MFCD22587390
  • Cas Number : 1395786-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart