LIF372153143 Life Chemicals Inc.

2-(1-naphthyloxy)propanoic acid

  • MDL number MFCD00021470
  • Cas Number : 13949-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart