LIF372153150 Life Chemicals Inc.

4-[(Z)-(2-amino-4-oxo-1,3-thiazol-5(4H)-ylidene)methyl]benzoic acid

  • MDL number MFCD05885486
  • Cas Number : 644996-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart