LIF372153151 Life Chemicals Inc.

Methyl 4-[(Z)-(2-amino-4-oxo-1,3-thiazol-5(4H)-ylidene)methyl]benzoate

  • MDL number MFCD01460314
  • Cas Number : 313226-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart