LIF372153153 Life Chemicals Inc.

5,6-dimethoxy-1,3-benzothiazol-2-amine

  • MDL number MFCD01663342
  • Cas Number : 6294-52-6
  • Molecular Weight : 210.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart