LIF372153161 Life Chemicals Inc.

5-Bromo-4-methylnicotinic acid

  • MDL number MFCD07439973
  • Cas Number : 677702-58-8
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart