LIF372153175 Life Chemicals Inc.

5-Bromo-[1,7]naphthyridine

  • MDL number MFCD09863862
  • Cas Number : 17965-76-3
  • Molecular Weight : 209.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart