LIF372153183 Life Chemicals Inc.

4-azepan-1-ylpyridine-2-carboxylic acid

  • MDL number MFCD11181443
  • Cas Number : 1094408-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart