LIF372153187 Life Chemicals Inc.

4-[4-(tert-butoxycarbonyl)piperazin-1-yl]pyridine-2-carboxylic acid

  • MDL number MFCD12547236
  • Cas Number : 1211595-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart