LIF372153192 Life Chemicals Inc.

6-bromo-5-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine

  • MDL number MFCD12024531
  • Cas Number : 1150617-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart