LIF372153195 Life Chemicals Inc.

6-bromo-2-(4-methylpiperazin-1-yl)pyridin-3-amine

  • MDL number MFCD20486711
  • Cas Number : 220499-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart