LIF372153196 Life Chemicals Inc.

6-bromo-1,2,3,4-tetrahydro-1,5-naphthyridine

  • MDL number MFCD21091457
  • Cas Number : 1219022-46-4
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart