LIF372153199 Life Chemicals Inc.

5,6,7,8-tetrahydro-1,8-naphthyridine-2-carboxylic acid

  • MDL number MFCD06738877
  • Cas Number : 885278-22-8
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart