LIF372153200 Life Chemicals Inc.

1,2,3,4-tetrahydro-1,6-naphthyridine-8-carboxylic acid

  • MDL number MFCD21091455
  • Cas Number : 1219022-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart