LIF372153206 Life Chemicals Inc.

7-Chloro-3-methyl-3H-[1,2,3]triazolo[4,5-d]pyrimidine

  • MDL number MFCD07644656
  • Cas Number : 21323-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart