LIF372153208 Life Chemicals Inc.

2-(4-fluorophenyl)imidazo[1,2-{a}]pyridin-3-amine

  • MDL number MFCD02658343
  • Cas Number : 326883-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart