LIF372153211 Life Chemicals Inc.

2-phenylimidazo[1,2-a]pyrimidin-3-amine

  • MDL number MFCD07021267
  • Cas Number : 89185-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart