LIF372153217 Life Chemicals Inc.

3-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-methyl-1{H}-pyrazole-5-carboxylic acid

  • MDL number MFCD16652622
  • Cas Number : 1177272-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart