LIF372153221 Life Chemicals Inc.

6-ethoxypyridazine-3-carboxylic acid

  • MDL number MFCD12152589
  • Cas Number : 142054-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart