LIF372153241 Life Chemicals Inc.

3-biphenyl-4-yl-1-methyl-1{H}-pyrazole-5-carboxylic acid

  • MDL number MFCD16652628
  • Cas Number : 1177282-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart