LIF372153257 Life Chemicals Inc.

1-methyl-3-(1-naphthyl)-1{H}-pyrazole-5-carboxylic acid

  • MDL number MFCD16652635
  • Cas Number : 1177347-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart