LIF372153262 Life Chemicals Inc.

(pyridazin-3-ylthio)acetic acid

  • MDL number MFCD16652639
  • Cas Number : 794574-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart