LIF372153270 Life Chemicals Inc.

[(6-phenylpyrimidin-4-yl)thio]acetic acid

  • MDL number MFCD09261754
  • Cas Number : 927998-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart