LIF372153277 Life Chemicals Inc.

[3-(2-furyl)-6-oxopyridazin-1(6{H})-yl]acetic acid

  • MDL number MFCD16652644
  • Cas Number : 1105192-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart