LIF372153285 Life Chemicals Inc.

4-amino-6-(4-methoxyphenyl)pyridazin-3(2{H})-one

  • MDL number MFCD08052636
  • Cas Number : 893760-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart