LIF372153291 Life Chemicals Inc.

4-amino-6-(2-thienyl)pyridazin-3(2{H})-one

  • MDL number MFCD10009040
  • Cas Number : 1018299-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart