LIF372153298 Life Chemicals Inc.

ethyl 4-hydroxy-6-oxo-1-phenyl-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00243731
  • Cas Number : 339030-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart