LIF372153303 Life Chemicals Inc.

4-methoxy-6-oxo-1-phenyl-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD16652652
  • Cas Number : 1105193-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart