LIF372153331 Life Chemicals Inc.

2-(2-fluorophenoxy)propanoic acid

  • MDL number MFCD01931911
  • Cas Number : 17088-71-0
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart