LIF372153336 Life Chemicals Inc.

(4-methyl-6-oxopyrimidin-1(6{H})-yl)acetic acid

  • MDL number MFCD12402755
  • Cas Number : 1105193-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart