LIF372153341 Life Chemicals Inc.

2,4-dioxo-3-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid

  • MDL number MFCD16652661
  • Cas Number : 37804-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart