LIF372153371 Life Chemicals Inc.

6-(2,3-dihydro-1,4-benzodioxin-6-yl)pyridazin-3-ol

  • MDL number MFCD12530380
  • Cas Number : 54558-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart