LIF372153379 Life Chemicals Inc.

6-(4-ethylphenyl)pyridazin-3-ol

  • MDL number MFCD02676711
  • Cas Number : 55901-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart