LIF372153442 Life Chemicals Inc.

3-chloro-6-(thiophen-3-yl)pyridazine

  • MDL number MFCD16683705
  • Cas Number : 78784-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart