LIF372153449 Life Chemicals Inc.

6-(4-methylpiperazin-1-yl)pyridazin-3-ol

  • MDL number MFCD15194895
  • Cas Number : 344287-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart