LIF372153451 Life Chemicals Inc.

6-pyrrolidin-1-ylpyridazin-3-ol

  • MDL number MFCD14756662
  • Cas Number : 344296-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart