LIF372153456 Life Chemicals Inc.

2-(4-fluorophenyl)-8-methylimidazo[1,2-{a}]pyridin-3-amine

  • MDL number MFCD06015527
  • Cas Number : 89185-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart