LIF372153473 Life Chemicals Inc.

6-chloro-{N}-(4-fluorobenzyl)pyridazin-3-amine

  • MDL number MFCD07681959
  • Cas Number : 872102-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart