LIF372153495 Life Chemicals Inc.

6-isopropylpyrimidin-4-ol

  • MDL number MFCD09909771
  • Cas Number : 124703-81-7
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart