LIF372153513 Life Chemicals Inc.

4-chloro-6-pyridin-2-ylpyrimidine

  • MDL number MFCD09750195
  • Cas Number : 284050-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart