LIF372153520 Life Chemicals Inc.

6-(2-methoxyphenyl)-2-thioxo-2,3-dihydropyrimidin-4(1{H})-one

  • MDL number MFCD16652727
  • Cas Number : 886140-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart