LIF372153523 Life Chemicals Inc.

6-pyridin-2-yl-2-thioxo-2,3-dihydropyrimidin-4(1{H})-one

  • MDL number MFCD16652728
  • Cas Number : 64670-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart