LIF372153545 Life Chemicals Inc.

6-chloro-3-(3-methoxyphenyl)[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD11540514
  • Cas Number : 596825-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart