LIF372153569 Life Chemicals Inc.

3-cyclopropyl-6-piperazin-1-yl[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD16652738
  • Cas Number : 1204296-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart